missing translation for 'onlineSavingsMsg'
Learn More

INF2 Mouse anti-Human, CoraLite 488, Clone: 4A11A7, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech CL488-66910-100UL

 View more versions of this product

Product Code. 17609876

  • 450.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

INF2 Monoclonal antibody apecifically detetects INF2 antigen in Human Samples. It is validated for Immunofluorescence.
Specifications

Specifications

INF2
Monoclonal
1000 μg/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
INF2
INF2 fusion protein Ag17043
100 μL
64423
Store at -20°C. Avoid exposure to light.
IgG2a
Immunofluorescence
4A11A7
CoraLite 488
BC008756
Mouse
Protein A purification
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.