Learn More
Indigo Carmine Solution - Ph Eur Ref: 1045601, Fisher Chemical™
Product Code. 30695135 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30695135 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30695135 Supplier Fisher Chemical Supplier No. RP 102 100
  • ONLINE EXCLUSIVE PRICE
    63.50€
    51.03€ / 100mL
    Save 12.47€
    19% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Pharmacopoeia reagents for testing and monographs, produced under a GMP-compliant ISO 9001 quality management system.
    This Pharmacopoeia reagent is manufactured :
  • Using pharmacopoeia reagent quality components
  • Water meeting the relevant pharmacopoeia monograph requirement for Purified Water
  • Supplied with a Certificate of Analysis for Performance Test
  • Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO 17034
Specifications
Quantity 100 mL