missing translation for 'onlineSavingsMsg'
Learn More

Indapamide Lactose Adduct Impurity, TRC

Product Code. 30408700
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408700 10 mg 10mg
30403447 5 mg 5mg
30395894 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408700 Supplier Toronto Research Chemicals Supplier No. TRCI79694010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Indapamide Lactose Adduct Impurity
Analyte or Component Names Indapamide Lactose Adduct Impurity
Shipping Condition Room Temperature
Molecular Formula C28H36ClN3O13S
Synonym 4-chloro-3-(N-((2R,3R,4R,5S,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)sulfamoyl)-N-(2-methylindolin-1-yl)benzamide
InChI Formula InChI=1S/C28H36ClN3O13S/c1-12-8-13-4-2-3-5-16(13)32(12)30-26(40)14-6-7-15(29)19(9-14)46(41,42)31-27-23(38)22(37)25(18(11-34)43-27)45-28-24(39)21(36)20(35)17(10-33)44-28/h2-7,9,12,17-18,20-25,27-28,31,33-39H,8,10-11H2,1H3,(H,30,40)/t12?,17-,18-,20+,21+,22-,23-,24-,25-,27-,28+/m1/s1
Solution Type Neat
SMILES O=C(C1=CC=C(C(S(=O)(N[C@H]2[C@@H]([C@H]([C@@H]([C@H](O2)CO)O[C@H]3[C@@H]([C@H]([C@H]([C@H](O3)CO)O)O)O)O)O)=O)=C1)Cl)NN4C(CC5=C4C=CC=C5)C
Industry Type Pharmaceutical
Molecular Weight (g/mol) 690.11
Quantity 10 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.