missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin alpha 2/KPNA2 Antibody (C11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43400
This item is not returnable.
View return policy
Description
Importin alpha 2/KPNA2 Monoclonal antibody specifically detects Importin alpha 2/KPNA2 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Importin alpha 2/KPNA2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.01-2ug/mL, Immunohistochemistry 5-20 μg/mL, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunoprecipitation | |
| importin alpha 1, importin-alpha-P1, IPOA1, karyopherin alpha 2 (RAG cohort 1, importin alpha 1), Karyopherin subunit alpha-2, pendulin, QIP2importin alpha 2, RAG cohort 1, RAG cohort protein 1, RCH1importin subunit alpha-2, SRP1, SRP1alpha, SRP1-alpha | |
| Immunized with recombinant Human Importin alpha 2/KPNA2 (Uniprot#: P52292; Ala157∽Gln412) | |
| 100 μL | |
| Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Immunology, Stem Cell Markers | |
| 3838 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| C11 | |
| Unconjugated | |
| PBS, pH 7.4, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction