missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin alpha 2/KPNA2 Antibody (1E8B7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
205.00€ - 552.00€
Specifications
| Antigen | Importin alpha 2/KPNA2 |
|---|---|
| Clone | 1E8B7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18239600
|
Novus Biologicals
NBP2-52501 |
0.1 mg |
552.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656927
|
Novus Biologicals
NBP2-52501-0.025mg |
0.025 mg |
205.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Importin alpha 2/KPNA2 Monoclonal specifically detects Importin alpha 2/KPNA2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Importin alpha 2/KPNA2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Immunology, Stem Cell Markers | |
| PBS with 0.05% Sodium Azide | |
| 3838 | |
| Purified recombinant fragment of human KPNA2 (AA: 1-530) expressed in E. Coli. | |
| Primary | |
| 58 kDa |
| 1E8B7 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| importin alpha 1, importin-alpha-P1, IPOA1, karyopherin alpha 2 (RAG cohort 1, importin alpha 1), Karyopherin subunit alpha-2, pendulin, QIP2importin alpha 2, RAG cohort 1, RAG cohort protein 1, RCH1importin subunit alpha-2, SRP1, SRP1alpha, SRP1-alpha | |
| KPNA2 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title