missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Imiloxan hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
A moderately potent, but highly selective α2-adrenoceptor antagonist, the only reported selective antagonist at the α2B-adrenoceptor not having potent α1-adrenoceptor antagonist activity.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 280.75 |
| Purity | >98% |
| Molecular Weight (g/mol) | 280.752 |
| Chemical Name or Material | Imiloxan hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction