accessibility menu, dialog, popup

UserName

CAS RN 81167-22-8

CH3NNOOHCl

CAS RN 81167-22-8

IUPAC Name: 2-[(2,3-dihydro-1,4-benzodioxin-2-yl)methyl]-1-ethyl-1H-imidazole hydrochloride
Synonyms: imiloxan hcl
Molecular Weight (g/mol): 280.75
Molecular Formula: C14H17ClN2O2
InChi Key: ANDJPJNFVJXEKX-UHFFFAOYNA-N
SMILES: Cl.CCN1C=CN=C1CC1COC2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results