Learn More
Imazamethabenz-methyl 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30316775
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 30316775
Brand: Dr. Ehrenstorfer DREXA14281500AL
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Imazamethabenz-methyl
CAS 81405-85-8
Analyte or Component Names Imazamethabenz-methyl
Shipping Condition Room Temperature
Molecular Formula C16 H20 N2 O3
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Benzoic acid, 2-[4,5-dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl]-4(or 5)-methyl-, methyl ester (9CI, ACI), AC 222293, Assert, Assert 300, Dagger, Dagger G, Imazamethabenz methyl ester, Imazamethabenz-methyl
InChI Formula InChI=1S/2C16H20N2O3/c1-9(2)16(4)15(20)17-13(18-16)12-8-10(3)6-7-11(12)14(19)21-5;1-9(2)16(4)15(20)17-13(18-16)11-7-6-10(3)8-12(11)14(19)21-5/h2*6-9H,1-5H3,(H,17,18,20)
Solution Type Single Solution
SMILES COC(=O)c1ccc(C)cc1C2=NC(C)(C(C)C)C(=O)N2.COC(=O)c3cc(C)ccc3C4=NC(C)(C(C)C)C(=O)N4
IUPAC Name methyl 4-methyl-2-(4-methyl-5-oxo-4-propan-2-yl-1H-imidazol-2-yl)benzoate;methyl 5-methyl-2-(4-methyl-5-oxo-4-propan-2-yl-1H-imidazol-2-yl)benzoate
Solvent or Matrix Acetonitrile
Industry Type Environmental Testing
Molecular Weight (g/mol) 288.34
Quantity 1 mL
Formula Weight 288.1474 g/mol