missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ILK (Human) Recombinant Protein

Human ILK partial ORF ( AAH01554, 341 a.a. - 452 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00003611-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16106881

  • 349.00€ / 10µg

Estimated Shipment: 18-07-2023
to see stock.
Add to basket

Description

Description

  • Sequence: KFSFQCPGRMYAPAWVAPEALQKKPEDTNRRSADMWSFAVLLWELVTREVPFADLSNMEIGMKVALEGLRPTIPPGISPHVCKLMKICMNEDPAKRPKFDMIVPILEKMQDK
Specifications

Specifications

AAH01554
integrin-linked kinase
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ILK
GST
3611
Wheat germ expression system
10 μg
DKFZp686F1765, P59
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers