missing translation for 'onlineSavingsMsg'
Learn More

IL22RA2 Mouse anti-Human, Clone: 5G11A9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66190-1-IG-20UL

 View more versions of this product

Product Code. 16807845

  • 145.00€ / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IL22, a homolog of IL10, is secreted by T cells and binds to and signals through the class II cytokine receptor family (CRF) heterodimer IL22RA /IL10RB. IL22 induces the production of acute-phase reactants.
Specifications

Specifications

IL22RA2
Monoclonal
1.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q969J5
IL22RA2
IL22RA2 Fusion Protein Ag18682
20 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
5G11A9
Unconjugated
IL22RA2
CRF2 10, CRF2 S1, CRF2X, IL 22 receptor subunit alpha 2, IL 22BP, IL 22R alpha 2, IL 22RA2, IL22BP, IL22RA2, Interleukin 22 binding protein, ZcytoR16
Mouse
Protein A
RUO
116379
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.