missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IL1RAP (Human) Recombinant Protein
Human IL1RAP full-length ORF ( AAH53621.1, 1 a.a. - 356 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003556-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MTLLWCVVSLYFYGILQSDASERCDDWGLDTMRQIQVFEDEPARIKCPLFEHFLKFNYSTAHSAGLTLIWYWTRQDRDLEEPINFRLPENRISKEKDVLWFRPTLLNDTGNYTCMLRNTTYCSKVAFPLEVVQKDSCFNSPMKLPVHKLYIEYGIQRITCPNVDGYFPSSVKPTITWYMGCYKIQNFNNVIPEGMNLSFLIALISNNGNYTCVVTYPENGRTFHLTRTPTVKVVGSPKNAVPPVIHSPNDHVVYEKEPGEELLIPCTVYFSFLMDSRNEVWWTIDGKKPDDITIDVTINESISHSRTEDETRTQILSIKKVTSEDLKRSYVCHARSAKGEVAKAAKVKQKGNRCGQ
Specifications
AAH53621.1 | |
interleukin 1 receptor accessory protein | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL1RAP | |
GST |
3556 | |
Wheat germ expression system | |
25 μg | |
C3orf13, FLJ37788, IL-1RAcP, IL1R3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |