missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IL1RAP (Human) Recombinant Protein
Human IL1RAP full-length ORF ( AAH53621.1, 1 a.a. - 356 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | AAH53621.1 |
---|---|
Gene ID (Entrez) | 3556 |
Name | interleukin 1 receptor accessory protein |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16146741
|
Abnova™
H00003556-P01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16156741
|
Abnova™
H00003556-P01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
- Sequence: MTLLWCVVSLYFYGILQSDASERCDDWGLDTMRQIQVFEDEPARIKCPLFEHFLKFNYSTAHSAGLTLIWYWTRQDRDLEEPINFRLPENRISKEKDVLWFRPTLLNDTGNYTCMLRNTTYCSKVAFPLEVVQKDSCFNSPMKLPVHKLYIEYGIQRITCPNVDGYFPSSVKPTITWYMGCYKIQNFNNVIPEGMNLSFLIALISNNGNYTCVVTYPENGRTFHLTRTPTVKVVGSPKNAVPPVIHSPNDHVVYEKEPGEELLIPCTVYFSFLMDSRNEVWWTIDGKKPDDITIDVTINESISHSRTEDETRTQILSIKKVTSEDLKRSYVCHARSAKGEVAKAAKVKQKGNRCGQ
Specifications
AAH53621.1 | |
interleukin 1 receptor accessory protein | |
125% SDS-PAGE Stained with Coomassie Blue | |
C3orf13, FLJ37788, IL-1RAcP, IL1R3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
3556 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL1RAP | |
GST |