missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL17C Monoclonal Antibody (311523)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561135

Product Code. 30534816

  • 459.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Cytokine that plays a crucial role in innate immunity of the epithelium, including to intestinal bacterial pathogens, in an autocrine manner. Stimulates the production of antibacterial peptides and proinflammatory molecules for host defense by signaling through the NF-kappa-B and MAPK pathways. Acts synergically with IL22 in inducing the expression of antibacterial peptides, including S100A8, S100A9, REG3A and REG3G. Synergy is also observed with TNF and IL1B in inducing DEFB2 from keratinocytes. Depending on the type of insult, may have both protective and pathogenic properties, either by maintaining epithelial homeostasis after an inflammatory challenge or by promoting inflammatory phenotype. Enhanced IL17C/IL17RE signaling may also lead to greater susceptibility to autoimmune diseases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL17C
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q8K4C5
IL17C
E. coli-derived recombinant mouse IL-17C, His15-Gln194.
100 μg
Primary
Mouse
Antibody
IgG2a
Western Blot
311523
Unconjugated
IL17C
CX2; cytokine CX2; Il17c; IL-17C; IL-21; Interleukin; interleukin 17C; interleukin-17C; UNQ561/PRO1122
Rat
Protein A/G
RUO
234836
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.