Learn More
Abnova™ IL12RB1 Recombinant Protein
Human IL12RB1 full-length ORF recombinant protein with GST-tag at N-terminal
Brand: Abnova™ H00003594-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a type I transmembrane protein that belongs to the hemopoietin receptor superfamily. This protein binds to interleukin 12 (IL12) with a low affinity, and is thought to be a part of IL12 receptor complex. This protein forms a disulfide-linked oligomer, which is required for its IL12 binding activity. The co expression of this and IL12RB2 proteins was shown to lead to the formation of high-affinity IL12 binding sites and reconstitution of IL12 dependent signaling. The lack of expression of this gene was found to result in the immunodeficiency of patients with severe mycobacterial and Salmonella infections. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
- Theoretical MW (kDa): 67.65
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: MEPLVTWVVPLLFLFLLPRQGAACRTSECCFQDPPYPDADSGSASGPRDLRCYRISSDRYECSWQYEGPTAGVSHFLRCCLSSGRCCYFAAGSATRLQFSDQAGVSVLYTVTLWVESWARNQTEKSPEVTLQLYNSVKYEPPLGDIKVSKLAGQLRMEWETPDNQVGAEVQFRHRTPSSPWKLGDCGPQDDDTESCLCPLEMNVAQEFQLRRRRLGSQGSSWSKWSSPVCVPPENPPQPQVRFSVEQLGQDGRRRLTLKEQPTQLELPEGCQGLAPGTEVTYRLQLHMLSCPCKAKATRTLHLGKMPYLSGAAYNVAVISSNQFGPGLNQTWHIPADTHTDGMISAHCNLRLPDSRDSPASASRVAGITGICHHTRLILYF
Best use within three months from the date of receipt of this protein.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH29121 | |
Solution | |
3594 | |
IL12RB1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL12RB1 | |
Human | |
Yes |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
67.3 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MEPLVTWVVPLLFLFLLPRQGAACRTSECCFQDPPYPDADSGSASGPRDLRCYRISSDRYECSWQYEGPTAGVSHFLRCCLSSGRCCYFAAGSATRLQFSDQAGVSVLYTVTLWVESWARNQTEKSPEVTLQLYNSVKYEPPLGDIKVSKLAGQLRMEWETPDNQVGAEVQFRHRTPSSPWKLGDCGPQDDDTESCLCPLEMNVAQEFQLRRRRLGSQGSSWSKWSSPVCVPPENPPQPQVRFSVEQLGQDGRRRLTLKEQPTQLELPEGCQGLAPGTEVTYRLQLHMLSCPCKAKATRTLHLGKMPYLSGAAYNVAVISSNQFGPGLNQTWHIPADTHTDGMISAHCNLRLPDSRDSPASASRVAGITGICHHTRLILYF | |
CD212/IL-12R-BETA1/IL12RB/MGC34454 | |
IL12RB1 | |
Wheat Germ (in vitro) | |
GST |