missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL12RB1 Recombinant Human Monoclonal Antibody (7C11)
GREENER_CHOICE

Human Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA560734

Product Code. 30535182

  • 482.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IL12RB1 Recombinant Monoclonal Antibody for ELISA

The protein encoded by this gene is a type I transmembrane protein that belongs to the hemopoietin receptor superfamily. This protein binds to interleukin 12 with a low affinity, and is thought to be a part of IL12 receptor complex. This protein forms a disulfide-linked oligomer, which is required for its IL12 binding activity. The coexpression of this and IL12RB2 proteins was shown to lead to the formation of high-affinity IL12 binding sites and reconstitution of IL12 dependent signaling. The lack of expression of this gene was found to result in the immunodeficiency of patients with severe mycobacterial and Salmonella infections. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL12RB1
Recombinant Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
P42701
IL12RB1
Recombinant Human IL12RB1 protein.
100 μL
Primary
Human
Antibody
IgG1
ELISA
7C11
Unconjugated
IL12RB1
CD212; cluster of differentiation 212; IL-12 receptor beta component; IL-12 receptor subunit beta-1; IL12R; IL-12R subunit beta-1; IL-12R[b]; IL12RB; Il12rb1; IL-12RB1; IL-12R-BETA1; IL-12R-beta-1; IMD30; interleukin 12 receptor subunit beta 1; interleukin 12 receptor, beta 1; interleukin-12 receptor beta-1 chain; Interleukin-12 receptor beta-1 chain precursor (IL-12R-beta1) (Interleukin-12 receptor beta) (IL-12 receptor beta component); interleukin-12 receptor subunit beta-1; MGC34454
Human
Affinity chromatography
RUO
3594
-20°C or -80°C if preferred
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.