Learn More
IL-6R Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23457-1-AP-150UL
This item is not returnable.
View return policy
Description
Interleukin 6 receptor (IL6R), also known as CD126 (Cluster of Differentiation 126), is a type I cytokine receptor. This gene encodes a subunit of the interleukin 6 (IL6) receptor complex. Interleukin 6 is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in the immune response. The IL6 receptor is a protein complex consisting of this protein and interleukin 6 signal transducer (IL6ST/GP130/IL6-beta), a receptor subunit also shared by many other cytokines. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer. Alternatively spliced transcript variants encoding distinct isoforms have been reported. A pseudogene of this gene is found on chromosome 9.Specifications
| IL-6R | |
| Polyclonal | |
| Unconjugated | |
| IL6R | |
| CD126, gp80, IL 6 receptor subunit alpha, IL 6R 1, IL 6R alpha, IL 6R subunit alpha, IL 6RA, IL-6 R alpha, IL-6R, IL6RA, interleukin 6 receptor, Membrane glycoprotein 80 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16194, 24499, 3570 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08887, P22272, P22273 | |
| IL6R, Il6ra | |
| IL-6R Fusion Protein Ag18263 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |