missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL-4R alpha Antibody (MEDI2045), Novus Biologicals™
Human Monoclonal Antibody
213.00€ - 920.00€
Specifications
| Antigen | IL-4R alpha |
|---|---|
| Clone | MEDI2045 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30045784
|
Novus Biologicals™
NBP3-28409-1mg |
1 mg |
920.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30048217
|
Novus Biologicals™
NBP3-28409-100ug |
100 μg |
326.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30048027
|
Novus Biologicals™
NBP3-28409-50ug |
50 μg |
213.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-4R alpha Monoclonal antibody specifically detects IL-4R alpha in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| IL-4R alpha | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Asthma, B Cell Development and Differentiation Markers, Cancer, Immunology, Myeloid derived Suppressor Cell | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD124 antigen, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha | |
| IL-4Ra/ CD124 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| MEDI2045 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P24394 | |
| 3566 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title