missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-37/IL-1F7 Mouse anti-Human, Clone: B-P47, Novus Biologicals™
Mouse Monoclonal Antibody
736.00€
Specifications
| Antigen | IL-37/IL-1F7 |
|---|---|
| Clone | B-P47 |
| Concentration | 1 mg/ml |
| Dilution | ELISA |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18327054
|
Novus Biologicals
NBP3-18112-200UL |
200 μg |
N/A
|
N/A | |||||
|
18352315
|
Novus Biologicals
NBP3-18112-500UL |
500 μg |
736.00€
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-37/IL-1F7 Monoclonal antibody specifically detects IL-37/IL-1F7 in Human samples. It is validated for ELISASpecifications
| IL-37/IL-1F7 | |
| 1 mg/ml | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Cytokine Research | |
| PBS. Sterile-filtered through 0.22 um. | |
| 27178 | |
| IgG1 κ | |
| Affinity purified |
| B-P47 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FIL1, FIL1(ZETA), FIL1ZFIL1 zeta, IL-1F7IL-1 zeta, IL1H4IL1F7 (canonical product IL-1F7b), IL-1H4IL-1F7b (IL-1H4, IL-1H, IL-1RP1), IL-1RP1IL-1X protein, IL1RP1interleukin 1 family member 7, IL-1X, interleukin 1 family, member 7 (zeta), interleukin 1, zeta, interleukin-1 family member 7, Interleukin-1 homolog 4, interleukin-1 superfamily z, Interleukin-1 zeta, Interleukin-1-related protein | |
| Recombinant human IL-37/IL-1F7. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
| Primary | |
| Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title