missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL-31RA Antibody (nemolizumab) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
213.00€ - 920.00€
Specifications
| Antigen | IL-31RA |
|---|---|
| Clone | nemolizumab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30044654
|
Novus Biologicals™
NBP3-28357-50ug |
50 μg |
213.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045024
|
Novus Biologicals™
NBP3-28357-100ug |
100 μg |
349.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046850
|
Novus Biologicals™
NBP3-28357-1mg |
1 mg |
920.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-31RA Monoclonal antibody specifically detects IL-31RA in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| IL-31RA | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Apoptosis | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| class I cytokine receptor, CRL3Glmr, Cytokine receptor-like 3, GLMR, GLM-RMGC125346, GP130 like receptor, Gp130-like monocyte receptor, Gp130-like receptor, hGLM-R, IL-31 receptor subunit alpha, IL-31R subunit alpha, IL-31RAGPLCRL, IL-31R-alpha, interleukin 31 receptor A, interleukin-31 receptor subunit alpha, PRO21384, soluble type I cytokine receptor CRL3, ZcytoR17 | |
| IL-31Ra | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| nemolizumab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q8NI17 | |
| 133396 | |
| IgG2 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title