missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-23R Antibody (753317), Allophycocyanin/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB16861APCCY7
This item is not returnable.
View return policy
Description
IL-23R Monoclonal antibody specifically detects IL-23R in Mouse samples. It is validated for Flow Cytometry
Specifications
| IL-23R | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 753317 | |
| Flow Cytometry | |
| IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R, Gly24-Asp372, Accession # Q8K4B4 | |
| 0.1 mL | |
| Immunology | |
| 149233 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction