Learn More
IL-23R Antibody (218213) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB14001R
Product Code. 18558894
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
IL-23R Monoclonal antibody specifically detects IL-23R in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-23R
Monoclonal
Alexa Fluor 647
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot
218213
Western Blot
IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Mouse myeloma cell line NS0-derived recombinant human IL-23 R, Gly24-Ile354, Accession # Q5VWK5
0.1 mL
Immunology
149233
Store at 4°C in the dark.
IgG2b