missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-18/IL-1F4 Antibody (125-2H), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81040H
This item is not returnable.
View return policy
Description
IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| IL-18/IL-1F4 | |
| Monoclonal | |
| HRP | |
| PBS | |
| IL18 | |
| Fusion of splenocytes from mice immunized with human IL-18/IL-1F4, with SP2/O mouse myeloma cells. | |
| 0.1 mL | |
| Primary | |
| Recognizes the active form of human IL-18/IL-1F4. | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, ELISA | |
| 125-2H | |
| Western Blot, ELISA | |
| Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3606 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction