missing translation for 'onlineSavingsMsg'
Learn More

IL-1 RI Antibody (10E2), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-26202-50ul

 View more versions of this product

Product Code. 30036136

  • 343.00€ / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IL-1 RI Monoclonal antibody specifically detects IL-1 RI in Human samples. It is validated for Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-1 RI
Monoclonal
Unconjugated
50% Glycerol, 0.01M PBS, PH 7.4
Mouse
Affinity purified
RUO
Primary
Human
Purified
Flow Cytometry, ELISA
10E2
Flow Cytometry 1:50-1:200, ELISA
CD121 antigen-like family member A, CD121a antigen, IL1R, IL1R1, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80
Recombinant Human IL-1 RI protein [UniProt P14778]
50 μL
Cancer, Cytokine Research, Immunology, Innate Immunity, Phospho Specific
3554
Store at -20 to -70C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.