Learn More
R&D Systems™ IKK gamma Inhibitor Peptide Set
Product Code. 18780644 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18780644 2 mg 2mg
18114365 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18780644 Supplier R&D Systems™ Supplier No. NBP226504
  • ONLINE EXCLUSIVE PRICE
    621.00€
    586.95€ / 2mg
    Save 34.05€
    5% Off
 Restricted Item
Estimated Shipment: 13-11-2026
A Novus product, part of R&D Systems. Applications: In vitro assay, In vivo assay, Functional (Inhibition), Block/Neutralize
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 2 mg
Product Line Novus