missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ihfA Polyclonal Antibody

Product Code. 17978972 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17978972 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17978972 Supplier Invitrogen™ Supplier No. PA5144435

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ihfA Polyclonal Antibody for Western Blot, ELISA

This protein is one of the two subunits of integration host factor, a specific DNA-binding protein that functions in genetic recombination as well as in transcriptional and translational control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ihfA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene ihfA
Gene Accession No. P64393
Gene Alias himA
Gene Symbols ihfA
Host Species Rabbit
Immunogen Recombinant Neisseria meningitidis serogroup B Integration host factor subunit alpha(ihfA) (1-100aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 61280954
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.