Learn More
IgG4 Mouse anti-Human, Biotin, Clone: OTI4B12, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA700558
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
IgG4 Monoclonal antibody specifically detects IgG4 in Human samples. It is validated for ELISASpecifications
| IgG4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P01861 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3503 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| OTI4B12 | |
| Biotin | |
| IGHG4 | |
| IGHG4; Immunoglobulin G; immunoglobulin heavy constant gamma 4 (G4m marker); ImmunoglobulinG; ImmunoglobulinG4 | |
| Native Human IgG4 protein | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.