missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG2 Antibody (SR1938), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21994-100ul
This item is not returnable.
View return policy
Description
IgG2 Monoclonal antibody specifically detects IgG2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| IgG2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| SR1938 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Constant region of heavy chain of IgG2, immunoglobulin gamma 2 (Gm marker), immunoglobulin heavy constant gamma 2 (Gm marker) | |
| A synthesized peptide derived from human IgG2 (Uniprot #: P01859) | |
| 100 μg | |
| Immunology | |
| 3501 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction