Learn More
IgG(H+L) Donkey anti-Human, Biotin, Secondary Antibody, Proteintech
Product Code. 17680337 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
200 μL
1 mL
Unit Size:
1mL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17680337 1 mL 1mL
17690337 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17680337 Supplier Proteintech Supplier No. SA00004101ML
 Restricted Item
In Stock
Donkey anti-Human Affinipure Secondary Antibody
Specifications
Antigen IgG(H+L)
Applications ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry
Conjugate Biotin
Formulation 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Host Species Donkey
Purification Method The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Quantity 1 mL
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid