Learn More
IgG(H+L) Donkey anti-Goat, FITC, Secondary Antibody, Proteintech
Product Code. 17610337 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
200 μL
500 μL
Unit Size:
200µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17610337 500 μL 500µL
17600337 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17610337 Supplier Proteintech Supplier No. SA000033500UL
 Restricted Item
In Stock
Donkey anti-Goat Affinipure Secondary Antibody
Specifications
Antigen IgG(H+L)
Applications Immunofluorescence, Flow Cytometry
Conjugate FITC
Formulation 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Host Species Donkey
Purification Method The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Quantity 500 μL
Primary or Secondary Secondary
Target Species Goat
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid