Learn More
IgG Fc Goat anti-Rabbit, Polyclonal , Abnova™
Goat anti-rabbit IgG recognizes rabbit IgG F(c) fragment.
Brand: Abnova PAB9399.50mg
This item is not returnable.
View return policy
Specifications
| IgG Fc | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| This product is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. | |
| RUO | |
| No reaction was observed against rabbit IgG F(ab')2. | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at −20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Polyclonal | |
| Goat anti-rabbit IgG recognizes rabbit IgG F(c) fragment. This secondary antibody was purified using a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. | |
| In 10mM sodium phosphate, 150mM NaCl, pH 7.2 | |
| Rabbit IgG F(c) fragment | |
| 50 mg | |
| Secondary | |
| Rabbit | |
| Lyophilized |