missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Fc Goat anti-Rabbit, HRP, Polyclonal Antibody, MP Biomedicals
Goat Polyclonal Antibody
Brand: MP Biomedicals 0855690
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Rabbit samples. It is validated for Enzyme Immunoassay, Immunohistochemistry, and Western Blotting
Specifications
| IgG Fc | |
| Polyclonal | |
| HRP | |
| Affinity Purified | |
| Secondary | |
| Store unopened product at 2° to 8°C. Store opened material no longer than 4 weeks at 2° to 8°C. For long term storage, aliquot and store with 40% glycerol (v/v) at -20°C. |
| ELISA, Immunohistochemistry, Western Blot | |
| Total Protein: 0.4mg/mL | |
| Goat | |
| 2 mL | |
| Rabbit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction