missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG F(ab')2 Goat anti-Human, Polyclonal , Abnova™
Goat anti-human IgG recognizes human IgG F(ab')2 fragment.
Brand: Abnova PAB9301.50mg
This item is not returnable.
View return policy
Specifications
| IgG F(ab')2 | |
| Polyclonal | |
| Goat anti-human IgG recognizes human IgG F(ab')2 fragment. This secondary antibody was purified using a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. | |
| In 10mM sodium phosphate, 150mM NaCl, pH 7.2 | |
| Human IgG F(ab')2 fragment | |
| 50 mg | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at −20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| This product is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. | |
| RUO | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction