missing translation for 'onlineSavingsMsg'
Learn More

IgG Antibody (RMG07), Novus Biologicals™

Product Code. 18677956 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18677956 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18677956 Supplier Novus Biologicals Supplier No. NBP262018
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Goat Monoclonal Antibody

IgG Monoclonal antibody specifically detects IgG in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications ELISA
Classification Monoclonal
Clone RMG07
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 0.05ug/mL - 1ug/mL
Formulation 50% Glycerol/PBS, 1% BSA
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Goat
Immunogen Mouse IgG
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Epitope Tags, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3500
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.