missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IGF2 Recombinant Rabbit Monoclonal Antibody (1D6)
GREENER_CHOICE

Product Code. 30536017
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30536017 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30536017 Supplier Invitrogen™ Supplier No. MA560640

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

IGF2 Recombinant Monoclonal Antibody for ELISA

The IGFs are mitogenic, polypeptide growth factors that stimulate the proliferation and survival of various cell types, including muscle, bone, and cartilage tissue in vitro. IGFs are predominantly produced by the liver, although a variety of tissues produce the IGFs at distinctive times. The IGFs belong to the Insulin gene family, which also contains insulin and relaxin. The IGFs are similar to insulin by structure and function, but have a much higher growth-promoting activity than insulin. IGF-II expression is influenced by placenta lactogen, while IGF-I expression is regulated by growth hormone. Both IGF-I and IGF-II signal through the tyrosine kinase type I receptor (IGF-IR), but IGF-II can also signal through the IGF-II/Mannose-6-phosphate receptor. Mature IGFs are generated by proteolytic processing of inactive precursor proteins, which contain N-terminal and C-terminal propeptide regions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF2
Applications ELISA
Classification Recombinant Monoclonal
Clone 1D6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IGF2
Gene Accession No. P01344
Gene Alias AL033362; C11orf43; FLJ22066; FLJ44734; GRDF; H-IGF-2; IGF; Igf2; Igf-2; IGFII; Igf-II; Insulin like growth factor; insulin like growth factor 2; insulin-like growth factor 2; insulin-like growth factor 2 (somatomedin A); insulin-like growth factor II; insulin-like growth factor II (IGF-II); Insulin-like growth factor II (somatomedin A); Insulin-like growth factor II Ala-25 Del; insulin-like growth factor II isoform 2 variant 4 preprotein; insulin-like growth factor type 2; M6pr; Mpr; MSA; multiplication-stimulating activity; Multiplication-stimulating polypeptide; Peg2; PP1446; PP9974; preptin; RNIGF2; Somatomedin-A; T3M-11-derived growth factor
Gene Symbols IGF2
Host Species Rabbit
Immunogen A synthesized peptide derived from human IGF2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3481
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.