missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IGF-I/IGF-1 Antibody (xentuzumab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
213.00€ - 920.00€
Specifications
| Antigen | IGF-I/IGF-1 |
|---|---|
| Clone | xentuzumab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30046055
|
Novus Biologicals™
NBP3-28287-100ug |
100 μg |
326.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30048647
|
Novus Biologicals™
NBP3-28287-50ug |
50 μg |
213.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044763
|
Novus Biologicals™
NBP3-28287-1mg |
1 mg |
920.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGF-I/IGF-1 Monoclonal antibody specifically detects IGF-I/IGF-1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| IGF-I/IGF-1 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Biologically Active Proteins, Cell Cycle and Replication, Signal Transduction | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C | |
| IGF-1 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| xentuzumab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P05019 | |
| 3479 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title