missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ IgE (ε-Heavy Chain) Rabbit Polyclonal Antibody with BSA and azide

Product Code. 10394389 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10394389 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10394389 Supplier Epredia™ Supplier No. RB1435A

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

IgE (ε-Heavy Chain), Rb PAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Research Areas: Immunoglobulins; The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Rabbit

Clone: Polyclonal

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: IgE isolated from a pool of normal human sera

Molecular Weight: 50kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgE (epsilon-Heavy Chain)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified Ab with BSA and Azide
Host Species Rabbit
Immunogen IgE isolated from a pool of normal human sera
Quantity 1 mL
Regulatory Status IVD
Research Discipline Immunoglobulins
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.