Learn More
IgA (alpha Chain) Goat anti-Mouse , Polyclonal, Secondary Antibody, MP Biomedicals™
Goat Polyclonal Antibody
Brand: MP Biomedicals 0855478
Description
- Product is goat affinity purified antibody to mouse IgA (a chain) and buffer salts.
- The affinity purified antibody is prepared from the specific goat IgG fraction by solid phase absorption.
- This product is suitable for use as a precipitating antibody.
- It gives sharp precipitin lines in immunoelectrophoresis and immunodiffusion techniques.
- This product is suitable for conjugation with enzymes, radiolabels, fluorochromes or other markers, and for attachment to solid supports for use as an immunoadsorbant.
Specifications
| Immunohistochemistry | |
| 1.0mg/mL ( Protien) | |
| Product isgoat affinity purified antibody to mouse IgA (a chain) and buffer salts. | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| Liquid |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| 1 mg | |
| Secondary | |
| Store at 2-8°C. |