Learn More
IgA (alpha Chain) Goat anti-Mouse , Polyclonal, Secondary Antibody, MP Biomedicals™
Goat Polyclonal Antibody
Brand:  MP Biomedicals 0855478
Product Code. 11419984
  • 367.00€ / 1mg
Description
Description
  • Product is goat affinity purified antibody to mouse IgA (a chain) and buffer salts.
  • The affinity purified antibody is prepared from the specific goat IgG fraction by solid phase absorption.
  • This product is suitable for use as a precipitating antibody.
  • It gives sharp precipitin lines in immunoelectrophoresis and immunodiffusion techniques.
  • This product is suitable for conjugation with enzymes, radiolabels, fluorochromes or other markers, and for attachment to solid supports for use as an immunoadsorbant.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry
1.0mg/mL ( Protien)
Product isgoat affinity purified antibody to mouse IgA (a chain) and buffer salts.
Affinity Purified
RUO
Mouse
Liquid
Polyclonal
Unconjugated
Goat
1 mg
Secondary
Store at 2-8°C.