missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Ig Lambda C Domain/IGLC2 Antibody (5E12B9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27124-0.1ml
This item is not returnable.
View return policy
Description
Ig Lambda C Domain/IGLC2 Monoclonal antibody specifically detects Ig Lambda C Domain/IGLC2 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Ig Lambda C Domain/IGLC2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| IGLC, immunoglobulin lambda constant 2 (Kern-Oz- marker), MGC20392, MGC45681 | |
| Purified recombinant fragment of human Ig Lambda C Domain/IGLC2 (AA: 1-106) expressed in E. Coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 5E12B9 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 3538 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction