missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig gamma-4 Chain C Region, Rabbit anti-Human, Clone: IGHG4/2042R, Novus Biologicals™
Rabbit Monoclonal Antibody
300.00€ - 528.00€
Specifications
| Antigen | Ig gamma-4 Chain C Region |
|---|---|
| Clone | IGHG4/2042R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18214356
|
Novus Biologicals
NBP2-79716-20UG |
20 μg |
300.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18273998
|
Novus Biologicals
NBP2-79716-100UG |
100 μg |
528.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ig gamma-4 Chain C Region Monoclonal specifically detects Ig gamma-4 Chain C Region in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Ig gamma-4 Chain C Region | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P01861 | |
| 3503 | |
| Recombinant full-length human Ig gamma-4 Chain C Region protein | |
| Primary | |
| Store at 4°C. |
| IGHG4/2042R | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| IGHG4, mmunoglobulin heavy constant gamma 4, mmunoglobulin heavy constant gamma 4 (G4m marker) | |
| IGHG4 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title