missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig, λ1, λ2 and λ3 Light Chain, Clone: R26-46, Biotin, Secondary Antibody, BD
Description
The R26-46 antibody reacts specifically with mouse Igs bearing λ1, λ2, or λ3 light chains. It does not react with κ light chain or heavy chain. Detection of surface immunoglobulin on Ig λ chain-secreting hybridoma cells has bee
Host Species: Rat
Clone: R26-46
Isotype: IgG2a, κ
Species Reactivity: Mouse
Immunogen: Pooled Mouse Ig
ELISA
Specifications
Specifications
| Antigen | Ig, λ1, λ2 & λ3 Light Chain |
| Classification | Monoclonal |
| Clone | R26-46 |
| Concentration | 0.5mg/mL |
| Conjugate | Biotin |
| Host Species | Rat |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?