missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT88 Antibody (2B2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42984
This item is not returnable.
View return policy
Description
IFT88 Monoclonal antibody specifically detects IFT88 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| IFT88 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| 2B2 | |
| Western Blot 1:500 - 1:5000 Immunoprecipitation1:200 - 1:2000 Immunohistochemistry 1:20 - 1:200, ELISA | |
| D13S1056ETPR repeat protein 10, DAF19, hTg737, intraflagellar transport 88 homolog (Chlamydomonas), intraflagellar transport protein 88 homolog, MGC26259, polaris homolog, probe hTg737 (polycystic kidney disease, autosomal recessive), Recessive polycystic kidney disease protein Tg737 homolog, tetratricopeptide repeat domain 10, Tetratricopeptide repeat protein 10, TG737Tg737, TTC10 | |
| intraflagellar transport 88 homolog(Chlamydomonas) - (Uniprot# Q13099) | |
| 100 μg | |
| Cell Biology, Cell Cycle and Replication | |
| 8100 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction