Learn More
IFT20 Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
Product Code. 16841914 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16841914 20 μL 20µL
16831914 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16841914 Supplier Proteintech Supplier No. 136151AP20UL
Rabbit Polyclonal Antibody
IFT20 is part of intraflagellar transport (IFT) particles involved in ciliary process assembly. IFT20 may play a role in the trafficking of ciliary membrane proteins from the Golgi complex to the cilium.
Specifications
Antigen IFT20
Applications Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IFT20
Gene Accession No. Q61025, Q8IY31
Gene Alias hIFT20, IFT20
Gene Symbols IFT20
Host Species Rabbit
Immunogen IFT20 Fusion Protein Ag4521
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 480620, 55978, 90410
Target Species Canine, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG