missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IFI27 Antibody (CL14627), Novus Biologicals™

Mouse Monoclonal Antibody

433.00€ - 688.00€

Specifications

Antigen IFI27
Clone CL14627
Concentration 1 mg/mL
Dilution Immunohistochemistry 1:200 - 1:500
Applications Immunohistochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043689
View Documents
Novus Biologicals™
NBP3-26876-25ul
25 μL
433.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043329
View Documents
Novus Biologicals™
NBP3-26876-100ul
100 μL
688.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IFI27 Monoclonal antibody specifically detects IFI27 in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

IFI27
1 mg/mL
Immunohistochemistry
Unconjugated
Mouse
Angiogenesis, Cancer
PBS, pH 7.2, 40% glycerol
3429
IgG1
Protein A purified
CL14627
Immunohistochemistry 1:200 - 1:500
Monoclonal
Purified
RUO
Human
FAM14D, Interferon alpha-induced 11.5 kDa protein, interferon alpha-inducible protein 27, mitochondrial, interferon, alpha-inducible protein 27,2310061N23Rik, Interferon-stimulated gene 12a protein, ISG12, ISG12(a), ISG12A, P27
This antibody was developed against a Synthetic Peptide of P40305. Actual immunogen sequence is proprietary.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.