Learn More
IdU Rat anti-Chemical, Clone: OTI5E2, lyophilized, TrueMAB™
Rat Monoclonal Antibody
Brand: Origene Technologies CF190141
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Idu (Idoxuridine) is a nucleoside analogue, a modified form of deoxyuridine, similar enough to be incorporated into viral DNA replication, but the iodine atom added to the uracil component blocks base pairing. It is used only topically due to cardiotoxicity. It was synthesized by William Prusoff in the late 1950s.Specifications
| IdU | |
| Monoclonal | |
| Unconjugated | |
| IdU | |
| Iododeoxyuridine coupled to keyhole limpet hemocyanin. | |
| 100 μg | |
| Primary | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin) | |
| OTI5E2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Rat | |
| Affinity Chromatography | |
| RUO | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized | |
| Chemical |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.