missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ID3 Antibody (OTI8B3), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-70979DL594
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ID3 Monoclonal antibody specifically detects ID3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifica
| ID3 | |
| Monoclonal | |
| DyLight 594 | |
| BHLHB25, bHLHb251R21, Class B basic helix-loop-helix protein 25, DNA-binding protein inhibitor ID-3, HEIR1, HEIR-1, Helix-loop-helix protein HEIR-1, ID-like protein inhibitor HLH 1R21, Inhibitor of DNA binding 3, inhibitor of DNA binding 3, dominant negative helix-loop-helix protein | |
| Full length recombinant protein of human ID3 (NP_002158) produced in HEK293T cells. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Prostate Cancer | |
| 3399 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI8B3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto