Learn More
ICP-MS Tuning Solution 2: Ce, Co, Li, Mg, Tl, Y @ 10 ug/mL in 2% HNO3, VHG™
Product Code. 17962326
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962326 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17962326 Supplier VHG Labs Supplier No. VHGLAGTSTK2100
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
ICP-MS Tuning Solution 2: Ce, Co, Li, Mg, Tl, Y @ 10 ug/mL in 2% HNO3
TRUSTED_SUSTAINABILITY
Specifica
Chemical Name or Material Cerium/Cobalt/Lithium/Magnesium/Thallium/Yttrium
CAS 7440-45-1, 7440-48-4, 7439-93-2, 7439-95-4, 7440-28-0, 7440-65-5
Analyte or Component Names Cerium/Cobalt/Lithium/Magnesium/Thallium/Yttrium
Shipping Condition Room Temperature
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Cerium 10 μg/mL/Cobalt 10 μg/mL/Lithium 10 μg/mL/Magnesium 10 μg/mL/Thallium 10 μg/mL/Yttrium 10 μg/mL
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.