missing translation for 'onlineSavingsMsg'
Learn More

ICOS (CD278) Armenian Hamster, BB515, Clone: C398.4A, BD Horizon™

Armenian Hamster Monoclonal Antibody

106.00€ - 240.00€

Specifications

Antigen ICOS (CD278)
Clone C398.4A
Concentration 0.2mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15844579
View Documents
BD Biosciences
565881
25 μg
106.00€
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15834579
View Documents
BD Biosciences
565880
0.1 mg
240.00€
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat , Rhesus Ma samples. It is validated for Flow cytometry.

ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat , Rhesus Ma samples. It is validated for Flow cytometry.
Specifications

Specifications

ICOS (CD278)
0.2mg/mL
Monoclonal
Purified
RUO
Human, Mouse, Rat, Rhesus Monkey
ICOS; AILIM; H4; Inducible T-cell costimulator; CVID1; Ly115
IgG
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
C398.4A
Flow Cytometry
Brilliant Blue 515 (BB515)
Armenian Hamster
Cell Biology
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse D10.G4.1 T-cell clone
Primary
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.