Learn More
IBA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Product Code. 16897593 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16897593 150 μL 150µL
16807603 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16897593 Supplier Proteintech Supplier No. 109041AP150UL
Rabbit Polyclonal Antibody
AIF1 is induced by cytokines and interferon. It is thought to be involved in the negative regulation of growth of vascular smooth muscle cells, which contributes to the anti-inflammatory response to vessel wall trauma.
Specifications
Antigen IBA1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene AIF1
Gene Accession No. O70200, P55008
Gene Alias AIF 1, AIF1, G1, IBA1, IRT 1, Protein G1
Gene Symbols AIF1
Host Species Rabbit
Immunogen IBA1 Fusion Protein Ag1363
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11629, 199
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG