missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Helicobacter pylori Mouse anti-Helicobacter pylori, Clone: J354, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB13557.1mg
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against Helicobacter pylori.
- High specificity
- High reactivity
Specifications
| Helicobacter pylori | |
| Monoclonal | |
| Unconjugated | |
| ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| H. pylori | |
| Liquid |
| ELISA, Immunofluorescence, Lateral Flow, Western Blot | |
| J354 | |
| Mouse monoclonal antibody raised against Helicobacter pylori. | |
| In 10 mM PBS, pH 7.2 (0.09% sodium azide). | |
| Protein A purification | |
| RUO | |
| This antibody reacts with flagellin of Helicobacter pylori. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction