missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cryptosporidium parvum Mouse anti-Cryptosporidium parvum, Clone: J350, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB13512.1mg
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against Cryptosporidium parvum.
- High specificity
- High reactivity
Specifications
| Cryptosporidium parvum | |
| Monoclonal | |
| Unconjugated | |
| ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| C. parvum | |
| Liquid |
| ELISA, Immunofluorescence, Lateral Flow | |
| J350 | |
| Mouse monoclonal antibody raised against Cryptosporidium parvum. | |
| In 10 mM PBS, pH 7.2 (0.09% sodium azide). | |
| Protein A purification | |
| RUO | |
| This antibody reacts with intact Cryptosporidium parvum oocysts. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction